Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Otubain-2 Antibody (OTI11B3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273172MFV450
Description
Otubain-2 Monoclonal antibody specifically detects Otubain-2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
Otubain-2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
78990 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI11B3 | |
Flow Cytometry | |
C14orf137chromosome 14 open reading frame 137, Deubiquitinating enzyme OTUB2, EC 3.4.19.12, FLJ21916, MGC3102, OTB2otubain-2, OTU domain, ubiquitin aldehyde binding 2, OTU domain-containing ubiquitin aldehyde-binding protein 2, OTU2ubiquitin thioesterase OTUB2, Otubain-2, ubiquitin-specific protease otubain 2, Ubiquitin-specific-processing protease OTUB2 | |
Full length human recombinant protein of human OTUB2 (NP_075601) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction