Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598487 Supplier Invitrogen™ Supplier No. PA598487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OTUD3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Deubiquitinating enzyme that hydrolyzes 'Lys-6'- and 'Lys-11'-linked polyubiquitin. Also hydrolyzes heterotypic (mixed and branched) and homotypic chains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.3
Gene OTUD3
Gene Accession No. Q5T2D3
Gene Alias DUBA4; KIAA0459; OTU deubiquitinase 3; OTU domain containing 3; OTU domain-containing protein 3; OTUD3; RP11-460G22.1
Gene Symbols OTUD3
Host Species Rabbit
Immunogen Recombinant Human OTU domain-containing protein 3 protein (108-398AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23252
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.