Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTUD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP237690
Description
OTUD4 Polyclonal specifically detects OTUD4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
OTUD4 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
54726 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
XP_005263137.1 | |
OTUD4 | |
This antibody maps to a region between residue 1063 to 1113 of human OTU Domain Dontaining 4 using the numbering given in entry XP_005263137.1 (GeneID 54726). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction