Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUD4 Monoclonal Antibody (3B12G11)
GREENER_CHOICE

Catalog No. PIMA531812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531812 Supplier Invitrogen™ Supplier No. MA531812
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31812 has been tested in indirect ELISA.

OTUD4 alternatively spliced transcript variants have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD4
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3B12G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene OTUD4
Gene Accession No. Q01804
Gene Alias 4930431L18Rik; AI449692; D8Ertd69e; DUBA6; HIN1; HIN-1; HIV-1 induced protein HIN-1; HIV-1-induced protein HIN-1; HSHIN1; KIAA1046; mKIAA1046; OTU deubiquitinase 4; OTU domain containing 4; OTU domain-containing protein 4; Otud4; RGD1305606
Gene Symbols OTUD4
Host Species Mouse
Immunogen Purified recombinant fragment of human OTUD4 (AA: 815-1049) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307774, 54726
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.