Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUD4 Mouse anti-Human, Rat, Clone:3B12G11, Abnova™

Catalog No. 89333573 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-573 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-573 Supplier Abnova Corporation Supplier No. MAB17192
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human OTUD4.

Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells. [provided by RefSeq]

Specifications

Antigen OTUD4
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3B12G11
Conjugate Unconjugated
Description OTU domain containing 4
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene OTUD4
Gene Alias DKFZp434I0721, DUBA6, HIN1, HSHIN1, KIAA1046
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 815-1049 of human OTUD4 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 54726
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.