Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557743 Supplier Invitrogen™ Supplier No. PA557743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CGSVSTVDEF PEARGEHVHS LPEASVSSKP DEGRTEQSSQ TRKADTALAS IPPVAEGKAH PPTQILNRER ETVPVELEPK RTIQS Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.

OTUD4 alternatively spliced transcript variants have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTUD4
Gene Accession No. Q01804
Gene Alias 4930431L18Rik; AI449692; D8Ertd69e; DUBA6; HIN1; HIN-1; HIV-1 induced protein HIN-1; HIV-1-induced protein HIN-1; HSHIN1; KIAA1046; mKIAA1046; OTU deubiquitinase 4; OTU domain containing 4; OTU domain-containing protein 4; Otud4; RGD1305606
Gene Symbols OTUD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OTUD4. Recombinant protein control fragment (Product #RP-96721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54726
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.