Learn More
OTUD4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304606AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells (provided by RefSeq).Specifications
OTUD4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
B2RRE7, Q01804 | |
Otud4 | |
Between 1063 and 1113 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OTUD4 | |
DUBA6, HIN1, HSHIN1; HIV-1 induced protein HIN-1; HIV-1-induced protein HIN-1; OTU domain-containing protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54726, 73945 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.