Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582587 Supplier Invitrogen™ Supplier No. PA582587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAASSGLEEW TSRSPRQRSS ASSPEHPELH AELGMKPPSP GTVLALAKPP SPCAPGTSSQ FSAGADRATS PLVSLYPALE CRALIQQMSP SAFGLNDWDD DEILASVLAV SQQEYLD Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 99%.

This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTUD5
Gene Accession No. Q96G74
Gene Alias AA407879; AI553596; BB114028; deubiquinating enzyme A; deubiquitinating enzyme A; DUBA; DXImx46e; OTU deubiquitinase 5; OTU domain containing 5; OTU domain-containing protein 5; Otud5; Sfc7
Gene Symbols OTUD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OTUD5. Recombinant protein control fragment (Product #RP-91986).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55593
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.