Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUD6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544960 Supplier Invitrogen™ Supplier No. PA544960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KMNLENRPPR SSKAHRKRER MESEERERQE SIFQAEMSEH LAGFKREEEE Sequence homology: Human: 100%.

Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA2 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD6A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OTUD6A
Gene Accession No. Q7L8S5
Gene Alias DUBA2; DUBA-2; EG408193; Hin6; Hin-6 protease; HSHIN6; OTU deubiquitinase 6A; OTU domain containing 6A; OTU domain-containing protein 6A; OTUD6A; RGD1560855
Gene Symbols OTUD6A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human OTUD6A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139562
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.