Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTUD6A Polyclonal Antibody

OTUD6A Polyclonal Antibody
Supplier: Thermo Scientific PA5110066
Description
OTUD6A Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| OTUD6A | |
| Polyclonal | |
| Unconjugated | |
| OTUD6A | |
| DUBA2; DUBA-2; EG408193; Hin6; Hin-6 protease; HSHIN6; OTU deubiquitinase 6A; OTU domain containing 6A; OTU domain-containing protein 6A; OTUD6A; RGD1560855 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 139562, 302426, 408193 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.79 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6IE21, Q7L8S5 | |
| OTUD6A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of mouse OTUD6A (Q6IE21). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction