Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OTUD6B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555092
Description
Immunogen sequence: HCMYKAIEDQ LKEKDCALTV VALRSQTAEY MQSHVEDFLP FLTNPNTGDM YTPEEFQKYC EDIVNTA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.
Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA5 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
Specifications
OTUD6B | |
Polyclonal | |
Unconjugated | |
OTUD6B | |
2600013N14Rik; AU015433; CGI-77; Deubiquitinase OTUD6B; DUBA5; DUBA-5; ortholog of human CGI-77; OTU domain containing 6B; OTU domain-containing protein 6B; Otud6b; RGD1310024 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
297911, 51633, 72201 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8K2H2, Q8N6M0 | |
OTUD6B | |
Recombinant protein corresponding to Human OTUD6B. Recombinant protein control fragment (Product #RP-93834). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction