Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUD6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110067
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110067 Supplier Invitrogen™ Supplier No. PA5110067
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEAVLTEELD EEEQLLRRHR KEKKELQAKI QGMKNAVPKN DKKRRKQLTE DVAKLEKEME QKHREELEQL KLTTKENKID SVAVNISNLV LENQPPRISK AQKRREKKAA LEKEREERIA EAEIENLTGA RHMESEKLAQ ILAARQLEIK QIPSDGHCMY KAIEDQLKEK DCALTVVALR SQTAEYMQSH VEDFLPFLTN PNTGDMYTPE EFQKYCEDIV NTAAWGGQLE LRALSHILQT PIEIIQADSP PIIVGEEYSK KPLILVYMRH AYGLGEHYNS VTRLVNIVTE NCS.

Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA5 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD6B
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OTUD6B
Gene Accession No. Q8K2H2, Q8N6M0
Gene Alias 2600013N14Rik; AU015433; CGI-77; Deubiquitinase OTUD6B; DUBA5; DUBA-5; ortholog of human CGI-77; OTU domain containing 6B; OTU domain-containing protein 6B; Otud6b; RGD1310024
Gene Symbols OTUD6B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-293 of human OTUD6B (NP_057107.4).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297911, 51633, 72201
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.