Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUD7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560745 Supplier Invitrogen™ Supplier No. PA560745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLVHGKMGRA NSANGKNGDS AERGKEKKAK SRKGSKEESG ASASTSPSEK TTPSPTDKAA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Has deubiquitinating activity towards 'Lys-11'-linked polyubiquitin chains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD7A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTUD7A
Gene Accession No. Q8TE49
Gene Alias C15orf16; C16ORF15; cezanne 2; CEZANNE2; OTU deubiquitinase 7A; OTU domain containing 7A; OTU domain-containing 7A; OTU domain-containing protein 7A; OTUD7; OTUD7A; Zinc finger protein Cezanne 2
Gene Symbols OTUD7A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OTUD7A. Recombinant protein control fragment (Product #RP-95851).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161725
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.