Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTUD7B/Cezanne/ZA20D1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31774825UL
This item is not returnable.
View return policy
Description
OTUD7B/Cezanne/ZA20D1 Polyclonal antibody specifically detects OTUD7B/Cezanne/ZA20D1 in Human samples. It is validated for ImmunofluorescenceSpecifications
OTUD7B/Cezanne/ZA20D1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
cellular zinc finger NF-kappaB Cezanne, Cellular zinc finger NF-kappa-B protein, CEZANNEA20 domain containing 1, EC 3.4.19.12, OTU domain containing 7B, OTU domain-containing protein 7B, ZA20D1, Zinc finger A20 domain-containing protein 1, Zinc finger protein Cezanne | |
This antibody was developed against Recombinant Protein corresponding to amino acids: QGEGKFIFVGTLKMGHRHQYQEEMIQRYLSDAEERFLAEQKQKEAERKIMNGGIGGGPPPAKKPEPDA | |
25 μg | |
Mucosal Immunology | |
56957 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction