Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUD7B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 50555608 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-555-608 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-555-608 Supplier Proteintech Group Inc Supplier No. 166051AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Negative regulator of the non-canonical NF-kappa-B pathway that acts by mediating deubiquitination of TRAF3, an inhibitor of the NF-kappa-B pathway, thereby acting as a negative regulator of B-cell responses. In response to non-canonical NF- kappa-B stimuli, deubiquitinates ′Lys-48′-linked polyubiquitin chains of TRAF3, preventing TRAF3 proteolysis and over-activation of non-canonical NF-kappa-B. Negatively regulates mucosal immunity against infections. Mediates deubiquitination of EGFR. Has deubiquitinating activity toward ′Lys-11′, ′Lys-48′ or ′Lys-63′- linked polyubiquitin chains. In vitro, has preference for ′Lys- 11′-linked polyubiquitin chains; however such data are unsure in vivo. Hydrolyzes both linear and branched forms of polyubiquitin.

Specifications

Antigen OTUD7B
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OTUD7B
Gene Accession No. B2RUR8, Q6GQQ9
Gene Alias CEZANNE, OTU domain containing 7B, OTU7B, OTUD7B, ZA20D1, Zinc finger protein Cezanne
Gene Symbols Otud7b
Host Species Rabbit
Immunogen OTUD7B Fusion Protein Ag9788
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229603, 310677, 56957
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.