Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTX1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735392 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5392 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5392 Supplier Proteintech Group Inc Supplier No. 265951AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probably plays a role in the development of the brain and the sense organs. Can bind to the BCD target sequence (BTS): 5′-TCTAATCCC-3′.

Specifications

Antigen OTX1
Applications Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene OTX1
Gene Accession No. P32242
Gene Alias Homeobox protein OTX1, orthodenticle homeobox 1, Orthodenticle homolog 1, OTX1
Gene Symbols OTX1
Host Species Rabbit
Immunogen OTX1 Fusion Protein Ag24252
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5013
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.