Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTX1 Recombinant Rabbit Monoclonal Antibody (6H7J9)
GREENER_CHOICE

Catalog No. PIMA542732
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542732 Supplier Invitrogen™ Supplier No. MA542732
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: MMSYLKQPPY GMNGLGLAGP AMDLLHPSVG YPATPRKQRR ERTTFTRSQL DVLEALFAKT RYPDIFMREE VALKINLPES RVQVWFKNRR AKCRQQQQSG.

Probably plays a role in the development of the brain and the sense organs. Can bind to the BCD target sequence (BTS): 5'-TCTAATCCC-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTX1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6H7J9
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OTX1
Gene Accession No. P80205, Q63410
Gene Alias A730044F23Rik; Homeobox protein OTX1; jv; orthodenticle homeobox 1; orthodenticle homolog 1; OTX; Otx1; Otx-1; OTX1X
Gene Symbols OTX1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human OTX1 (NP_0553771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18423, 25646
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.