Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Otx2 Antibody (PCRP-OTX2-1E10), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314028AF532
This item is not returnable.
View return policy
Description
Otx2 Monoclonal antibody specifically detects Otx2 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| Otx2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
| Recombinant human full-lengthOtx2protein (Uniprot: P32243) | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers, Vision | |
| 5015 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-OTX2-1E10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction