Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Otx2 Antibody (PCRP-OTX2-1E10), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314028MFV450
Description
Otx2 Monoclonal antibody specifically detects Otx2 in Human samples. It is validated for Flow CytometrySpecifications
Otx2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-OTX2-1E10 | |
Flow Cytometry | |
homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
Recombinant human full-lengthOtx2protein (Uniprot: P32243) | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers, Vision | |
5015 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction