Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTX2 Monoclonal Antibody (1H12C4B5)
GREENER_CHOICE

Catalog No. PIMA515854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515854 Supplier Invitrogen™ Supplier No. MA515854
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15854 targets OTX2 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15854 immunogen is purified recombinant fragment of human OTX2 expressed in E. Coli. . MA5-15854 detects OTX2 which has a predicted molecular weight of approximately 32kDa.

OTX2 is a 289 amino acid containing protein encoding a member of the bicoid sub-family of homeodomain-containing transcription factors. It has two known alternative splice variants with distinct isoforms though presence of other isoforms cannot be excluded. The protein is an early marker of endodermal differentiation of embryonic stem cells and may play a role in the development of the brain and the sensory organs. The proteinmay bind to BCD target sequence (BTS): 5-'TCTAATCCC-3'. Defects in OTX2 are the cause of microphthalmia syndromic type 5 (MCOPS5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTX2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1H12C4B5
Concentration Conc.not determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene OTX2
Gene Accession No. P32243, P80206, Q64201
Gene Alias CPHD6; E130306E05Rik; homeobox Otx2; homeobox protein OTX2; MCOPS5; orthodenticle homeobox 2; orthodenticle homolog 2; OTX; OTX2
Gene Symbols OTX2
Host Species Mouse
Immunogen Purified recombinant fragment of human OTX2 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18424, 305858, 5015
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.