Learn More
OTX2 Mouse anti-Human, Clone: OTI3G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809627

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
OTX2 is a 289 amino acid containing protein encoding a member of the bicoid sub-family of homeodomain-containing transcription factors. It has two known alternative splice variants with distinct isoforms though presence of other isoforms cannot be excluded. The protein is an early marker of endodermal differentiation of embryonic stem cells and may play a role in the development of the brain and the sensory organs. The proteinmay bind to BCD target sequence (BTS): 5-′TCTAATCCC-3′. Defects in OTX2 are the cause of microphthalmia syndromic type 5 (MCOPS5).Specifications
OTX2 | |
Monoclonal | |
Unconjugated | |
OTX2 | |
CPHD6, MCOPS5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5015 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P32243 | |
OTX2 | |
Full length human recombit protein of human OTX2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.