Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Otx2 Rabbit anti-Human, Mouse, Rat, Clone: 6I3J0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Otx2 |
---|---|
Clone | 6I3J0 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Otx2 Monoclonal antibody specifically detects Otx2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Otx2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers, Vision | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5015 | |
IgG | |
Affinity purified |
6I3J0 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
A synthetic peptide corresponding to a sequence within amino acids 190-289 of human Otx2 (P32243). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title