Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ouabain
GREENER_CHOICESDP

Catalog No. 1076100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-761-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-761-00 Supplier R&D Systems™ Supplier No. 1076/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Na+/K+ ATPase inhibitor

Selective Na+, K+-ATPase inhibitor. Cardiotonic agent. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.08 μM).

Chemical Identifiers

CAS 630-60-4
Molecular Formula C29H44O12

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 584.66
Purity >95%
Molecular Weight (g/mol) 584.659
Chemical Name or Material Ouabain
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.