Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Out Of Service Tags
Click to view available options
Message:
Out of Service
Description
- Hang lockout tag when working on equipment and machinery
- Lockout tag is required to mark and identify the lockout situation
- Spaces on tag to write in lockout info
- Pliable 10-mil poly-based tags will not crack, distort, or shrink
- Tear-resistant, waterproof, pulp-free
- Ideal for indoor and short-term outdoor conditions
Specifications
Specifications
Material | Pulp-free Cardstock |
Message | Out of Service |
Thickness | 10 mil |
Waterproof | Yes |
Dimensions (L x W) | 5.75 x 3.25 in. |
Diameter (English) Hole | 0.38 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction