Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite Outdoor Flammable Cabinet
GREENER_CHOICE

Catalog No. 19033907 Shop All Justrite Safety Group Products
Click to view available options
Dimensions (D x W x H):
18 x 43 x 44 in.
18 x 43 x 65 in.
Door Style:
Manual-close, Hinged, Three-point lock, Locking
Self-close, Hinged, Three-point lock, Locking, Auto-close

Engineered to withstand the elements and potential impacts, this cabinet is OSHA, NFPA, and FM approved, ensuring your flammable chemicals are safely and securely stored outdoors.

  • Spill Slope shelves with fully welded sump
  • Ground Screw
  • Durable and Chemical - resistant powder coated paint
  • UV resistant
  • Vent with built in flash arrestors
  • Welded Shelf Hangers
  • Leak Proof Sump

Specifications

No. of Doors 2
Color Yellow
Door Style Self-close, Hinged, Three-point lock, Locking, Auto-close
Material Steel (18 ga)
Shelf Type Adjustable
Certifications/Compliance OSHA, NFPA, and FM approved
For Use With (Application) Safe outdoor storage of flammable liquids to avoid risk of potential fire hazards and chemical spills in industries like chemical manufacturing, petrochemical and laboratories
Product Type Flammables Storage Cabinet
Weight (Metric) 87.4 kg
Description 30 gal.
Dimensions (D x W x H) Interior 14.6 x 39.5 x 39.6 in.
Weight (English) 192.7 lb.
Dimensions (D x W x H) 18 x 43 x 44 in.
No. of Shelves 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.