Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OV-6 Antibody (OV-6), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2020APCCY7
Description
OV-6 Monoclonal antibody specifically detects OV-6 in Human, Rat samples. It is validated for Flow CytometrySpecifications
OV-6 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Carcinogen-treated rat nodular hepatocytes | |
0.1 mL | |
Primary | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
OV-6 | |
Flow Cytometry | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction