Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), Brilliant Ultra Violet™ 615, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$215.00 - $437.00
Specifications
Antigen | OVA257-264 (SIINFEKL) peptide bound to H-2Kb |
---|---|
Clone | eBio25-D1.16 (25-D1.16) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
36-657-4380
![]() |
Invitrogen™
366574380 |
25 μg |
Each for $215.00
|
|
|||||
36-657-4382
![]() |
Invitrogen™
366574382 |
100 μg |
Each for $437.00
|
|
|||||
Description
OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow CytometrySpecifications
OVA257-264 (SIINFEKL) peptide bound to H-2Kb | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
IgG1 κ | |
Affinity chromatography |
eBio25-D1.16 (25-D1.16) | |
Flow Cytometry | |
Brilliant Ultraviolet 615 | |
Mouse | |
Mouse | |
H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title