Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oval Cell Marker Antibody (MIC1-1C3), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP118961MFV450
Description
Oval Cell Marker Monoclonal antibody specifically detects Oval Cell Marker in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Oval Cell Marker | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Nonparenchymal cells from DDC-treated mice | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
MIC1-1C3 | |
Flow Cytometry | |
Rat | |
Protein G purified | |
RUO | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction