Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oval Cell Marker Antibody (MIC1-1C3) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$243.50 - $562.50

Specifications

Antigen Oval Cell Marker
Clone MIC1-1C3
Concentration 1.0 mg/mL
Dilution Flow Cytometry 1:50 - 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature, Immunohistochemistry-Frozen 1:100, CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP118961SS
SDP
View Documents
Novus Biologicals
NBP118961SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NBP118961
SDP
View Documents
Novus Biologicals
NBP118961
0.1 mL
Each for $562.50
Only null left
Add to Cart
 
Description

Description

Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications

Specifications

Oval Cell Marker
1.0 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rat
Hepatic Stem Cell Markers, Stem Cell Markers
PBS with 0.09% Sodium Azide
Nonparenchymal cells from DDC-treated mice
Primary
Store at 4C. Do not freeze.
MIC1-1C3
Flow Cytometry 1:50 - 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature, Immunohistochemistry-Frozen 1:100, CyTOF-ready
Monoclonal
Supernatant
RUO
Human, Mouse
macrophage inhibitory cytokine-1
IgG2a
Protein G purified
Specific for hepatic proliferating duct cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.