Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oval Cell Marker Antibody (OC2-2F3), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP118964AF700
Description
Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Oval Cell Marker | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rat | |
Tissue culture supernatant | |
Primary | |
Mouse | |
Supernatant |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
OC2-2F3 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
macrophage inhibitory cytokine-1 | |
Nonparenchymal cells from DDC-treated mice | |
0.1 mL | |
Specific for proliferating hepatic duct cells. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction