Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oval Cell Marker Antibody (OC2-3C7), Novus Biologicals™
SDP

Rat Monoclonal Antibody has been used in 2 publications

$206.50 - $499.50

Specifications

Antigen Oval Cell Marker
Clone OC2-3C7
Concentration 1 mg/mL
Dilution Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Frozen 1:100
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP118970SS
SDP
View Documents
Novus Biologicals
NBP118970SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP118970
SDP
View Documents
Novus Biologicals
NBP118970
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

Oval Cell Marker
1 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Rat
Hepatic Stem Cell Markers, Stem Cell Markers
Tissue culture supernatant with 0.09% Sodium Azide
Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Primary
Store at 4C. Do not freeze.
OC2-3C7
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Frozen 1:100
Monoclonal
Supernatant
RUO
Mouse
macrophage inhibitory cytokine-1
IgG2a
Tissue culture supernatant
Specific for both proliferating and normal hepatic periductal cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.