Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oval Cell Marker Antibody (OC2-6E10), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP118981APCCY7
Description
Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry.Specifications
Oval Cell Marker | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
Nonparenchymal cells from DDC-treated mice to induce the oval cell response. | |
0.1 mL | |
Specific for proliferating hepatic periductal cells. | |
Store at 4°C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
OC2-6E10 | |
Flow Cytometry | |
Rat | |
Tissue culture supernatant | |
Primary | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction