Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oval Cell Marker Antibody (OC2-6E10), Novus Biologicals™
SDP

Catalog No. p-4984959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118981SS 0.025 mL
NBP118981 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP118981SS Supplier Novus Biologicals Supplier No. NBP118981SS
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 2 publications

Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Oval Cell Marker
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OC2-6E10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunohistochemistry-Frozen 1:100
Formulation Tissue culture supernatant with 0.09% Sodium Azide
Gene Alias macrophage inhibitory cytokine-1
Host Species Rat
Immunogen Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Purification Method Tissue culture supernatant
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Hepatic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Specific for proliferating hepatic periductal cells.
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.