Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Oval Cell Marker Monoclonal Antibody (OC2-1C6)
GREENER_CHOICE

Catalog No. PIMA516137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516137 Supplier Invitrogen™ Supplier No. MA516137
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Oval Cell Marker Monoclonal Antibody for Western Blot, IHC (F), Flow

Facultative liver stem cell activation was originally described in the rat, where chemical damage and partial hepatectomy caused the emergence of a new cell type, the oval cell. These cells are thought to be bipotential progenitors capable of yielding both hepatocytes and bile ducts. To properly determine the molecular regulation of the progenitor process, appropriate markers are needed to identify and isolate cells that emerge during the oval cell response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oval Cell Marker
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone OC2-1C6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.09% sodium azide
Host Species Rat
Immunogen Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.