Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ovalbumin Antibody (2C6), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062809JF549
Description
Ovalbumin Monoclonal antibody specifically detects Ovalbumin in Chicken samples. It is validated for ELISASpecifications
Ovalbumin | |
Monoclonal | |
Janelia Fluor 549 | |
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8 | |
Spleen cells from Ovalbumin immunized Balb/c mice were fused with cells of the mouse myeloma, P3U1 | |
0.1 mL | |
Primary | |
Chicken | |
Purified |
ELISA | |
2C6 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
396058 | |
Store at 4°C in the dark. | |
IgE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction