Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Ovalbumin (Egg White)

Catalog No. 89955415 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 g

Used for SDS-PAGE

#REF!

Specifications

Accession Number P01012
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 396058
Molecular Weight (g/mol) 45kDa
Name Ovalbumin (Egg White)
Preparation Method Native protein chromatographically purified from Egg White
Quantity 1 g
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name SERPINB14
Gene Symbol SERPINB14
Assays Conjugo-immuno determinations; Drug and pharmaceutical processing.
Species Chicken
Protein Tag None
Expression System Native protein chromatographically purified from Egg White
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.