Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Ovalbumin (Egg White)
Click to view available options
Quantity:
1 g
Description
#REF!
Specifications
Specifications
Accession Number | P01012 |
For Use With (Application) | SDS-PAGE |
Gene ID (Entrez) | 396058 |
Molecular Weight (g/mol) | 45kDa |
Name | Ovalbumin (Egg White) |
Preparation Method | Native protein purified from Egg White |
Quantity | 1 g |
Storage Requirements | Store at 4°C. |
Regulatory Status | RUO |
Common Name | SERPINB14 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction