Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ovalbumin (Egg White) Rabbit anti-Chicken, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS81825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS81825UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS81825UL Supplier MilliporeSigma Supplier No. ABS81825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Ovalbumin (Egg White) Antibody, Cat. No. ABS818, is a rabbit polyclonal antibody that detects chicken egg ovalbumin and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ovalbumin (Egg White)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Allergen Gal d II;Egg albumin;Plakalbumin
Gene Symbols SERPINB14
Host Species Rabbit
Immunogen Full-length purified chicken egg ovalbumin.
Molecular Weight of Antigen ∽43 kDa observed; 42.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects chicken egg ovalbumin.
Target Species Chicken
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.