Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ovalbumin Monoclonal Antibody (2C6)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA180396
Description
Mouse anti Ovalbumin antibody, clone 2C6 recognizes ovalbumin (OVA). The antibody is suitable for use as a mouse IgE standard in ELISA assays (Hamadaet al. 2003).
Chicken ovalbumin is the major protein in the white of the egg (and a favorite antigen in immunological research). Egg white contains a variety of proteins including ovalbumin, conalbumin, ovomucoi and lysozyme. It belonds to the serpin family and the Ov serpin subfamily. Ovalbumin can cause an allergic reaction in humans. Ovalbumin has been implicated in the development of the egg shell. Immunohistochemistry revealed that ovalbumin is found only in the mammillary bodies of decalcified shell, and is not distributed throughout the shell matrix. These results indicate that ovalbumin is present during the initial phase of shell formation and becomes incorporated into the protein matrix of the mammillary bodies. However, it is not yet clear whether ovalbumin at this site plays a specific role in shell mineralization.
Specifications
Ovalbumin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01012 | |
OVAL | |
Ovalbumin. | |
100 μg | |
Primary | |
Chicken | |
Antibody | |
IgE |
ELISA | |
2C6 | |
Unconjugated | |
OVAL | |
Allergen Gal d II; clade (ovalbumin); egg albumin; Gal d 2; member 14; OVA; OVAL; Ovalbumin; ovalbumin (SERPINB14); plakalbumin; serpin peptidase inhibitor; SERPINB14; unnamed protein product | |
Mouse | |
Purified | |
RUO | |
396058 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction