Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ovalbumin Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA1196HRP
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1196HRP 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1196HRP Supplier Invitrogen™ Supplier No. PA1196HRP
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

By Western blot, PA1-196-HRP detects ovalbumin at ~45kD. Multiple bands may be observed, which are likely due to the presence of multiple glycosylated and/or aggregated species of ovalbumin within a particular protein preparation.

Chicken ovalbumin is the major protein in the 'white' of the egg (and a favorite antigen in immunological research). Egg white contains a variety of proteins including ovalbumin, conalbumin, ovomucoi and lysozyme. It belonds to the serpin family and the Ov serpin subfamily. Ovalbumin can cause an allergic reaction in humans. Ovalbumin has been implicated in the development of the egg shell. Immunohistochemistry revealed that ovalbumin is found only in the mammillary bodies of decalcified shell, and is not distributed throughout the shell matrix. These results indicate that ovalbumin is present during the initial phase of shell formation and becomes incorporated into the protein matrix of the mammillary bodies. However, it is not yet clear whether ovalbumin at this site plays a specific role in shell mineralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ovalbumin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with proprietary stabilizer and 0.07% Kathon
Gene OVAL
Gene Accession No. P01012
Gene Alias Allergen Gal d II; clade (ovalbumin); egg albumin; Gal d 2; member 14; OVA; OVAL; Ovalbumin; ovalbumin (SERPINB14); plakalbumin; serpin peptidase inhibitor; SERPINB14; unnamed protein product
Gene Symbols OVAL
Host Species Rabbit
Immunogen Ovalbumin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396058
Target Species Chicken
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.