Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ovalbumin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597525
Description
Ovalbumin Polyclonal Antibody for Western Blot, ELISA
Chicken ovalbumin is the major protein in the white of the egg (and a favorite antigen in immunological research). Egg white contains a variety of proteins including ovalbumin, conalbumin, ovomucoi and lysozyme. It belonds to the serpin family and the Ov serpin subfamily. Ovalbumin can cause an allergic reaction in humans. Ovalbumin has been implicated in the development of the egg shell. Immunohistochemistry revealed that ovalbumin is found only in the mammillary bodies of decalcified shell, and is not distributed throughout the shell matrix. These results indicate that ovalbumin is present during the initial phase of shell formation and becomes incorporated into the protein matrix of the mammillary bodies. However, it is not yet clear whether ovalbumin at this site plays a specific role in shell mineralization.
Specifications
Ovalbumin | |
Polyclonal | |
Unconjugated | |
OVAL | |
Allergen Gal d II; clade (ovalbumin); egg albumin; Gal d 2; member 14; OVA; OVAL; Ovalbumin; ovalbumin (SERPINB14); plakalbumin; serpin peptidase inhibitor; SERPINB14; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
396058 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P01012 | |
OVAL | |
Chicken Ovalbumin (Native Protein). | |
100 μg | |
Primary | |
Chicken | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction