Learn More
VistaLab™ Technologies Ovation™ 12-Channel Pipetters PROMO
SDP
Catalog No. 22147102TI Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
2 to 125 μL
25 to 850 μL
5 to 250 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-147-102TI 5 to 250 μL
22-147-101TI 2 to 125 μL
22-147-103TI 25 to 850 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-147-102TI Supplier VistaLab™ Technologies Supplier No. 10600250TI
Add to Cart
Add to Cart
Straightforward, simple programming coupled with reliable, consistent accuracy and precision.
  • Ergonomically correct, comfort-driven design with electronic functions for repetitive procedures
  • Feature a tip acquisition system that provides a positive feedback signal to indicate every tip is securely seated and sealed; no manual manipulations needed
  • Upon completion of pipetting, all tips are discarded with the push of a button
  • Equipped with Multiple Dispense, Pipet, Reverse Pipet, Mix and Serial Dilute operations and liquid handling functions with recall of stored volume settings
  • Available in both left- and right-handed models with suitably placed LCD screen
  • For precision liquid dispensing from 0.5 to 850μL
Specifications
Accuracy ±0.8% at 250 μL, ±1% at 125 μL, ±2.5% (at 25 μL)
Compatible Tips Small tip
Color Blue
Model Right Handed
Number of Channels 12
Product Line Technologies Ovation
Type Electronic Pipette
Volume (Metric) 5 to 250 μL
Display LCD
Finger Style Ergonomic
Increments 1 μL
Warranty 1 Year