Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OVCA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | OVCA1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
OVCA1 Polyclonal specifically detects OVCA1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
OVCA1 | |
Polyclonal | |
Rabbit | |
Human | |
1801 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EAVVYLGDGRFHLESVMIANPNVPAYRYDPYSKVLSREHYDHQRMQAARQEAIATARSAK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Diphthamide biosynthesis protein 2 homolog-like 1, Diphthamide biosynthesis protein 2-like, diptheria toxin resistance protein required for diphthamide biosynthesis(Saccharomyces)-like 1, diptheria toxin resistance protein required for diphthamide biosynthesis-like 1, diptheria toxin resistance protein required for diphthamide biosynthesis-like 1(S. cerevisiae), DPH1 homolog, DPH1 homolog (S. cerevisiae), DPH2L, DPH2L1diphthamide biosynthesis protein 1, DPH2-like 1, DPH-like 1, DPH-like 1 (S. cerevisiae), FLJ33211, hsDph1, Ovarian cancer-associated gene 1 protein, ovarian tumor suppressor candidate 1, OVCA1candidate tumor suppressor in ovarian cancer 1 | |
DPH1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title