Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OVCA1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP156693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP156693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP156693 Supplier Novus Biologicals Supplier No. NBP156693
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OVCA1 Polyclonal specifically detects OVCA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen OVCA1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9BZG8
Gene Alias Diphthamide biosynthesis protein 2 homolog-like 1, Diphthamide biosynthesis protein 2-like, diptheria toxin resistance protein required for diphthamide biosynthesis(Saccharomyces)-like 1, diptheria toxin resistance protein required for diphthamide biosynthesis-like 1, diptheria toxin resistance protein required for diphthamide biosynthesis-like 1(S. cerevisiae), DPH1 homolog, DPH1 homolog (S. cerevisiae), DPH2L, DPH2L1diphthamide biosynthesis protein 1, DPH2-like 1, DPH-like 1, DPH-like 1 (S. cerevisiae), FLJ33211, hsDph1, Ovarian cancer-associated gene 1 protein, ovarian tumor suppressor candidate 1, OVCA1candidate tumor suppressor in ovarian cancer 1
Gene Symbols DPH1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to DPH1(DPH1 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of DPH1. Peptide sequence RMQAARQEAIATARSAKSWGLILGTLGRQGSPKILEHLESRLRALGLSFV.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1801
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Xenopus: 92%; Rat: 92%; Zebrafish: 92%; Chicken: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.