Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OVCA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569810 Supplier Invitrogen™ Supplier No. PA569810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 92%; Zebrafish: 85%.

Diphthamide is a unique posttranslationally modified histidine found only in translation elongation factor-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OVCA1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DPH1
Gene Accession No. Q9BZG8
Gene Alias 2-(3-amino-3-carboxypropyl)histidine synthase subunit 1; 2310011M22Rik; 4930488F09Rik; AW551873; candidate tumor suppressor in ovarian cancer 1; DEDSSH; diphthamide biosynthesis 1; diphthamide biosynthesis protein 1; diphthamide biosynthesis protein 1; 2-(3-amino-3-carboxypropyl)histidine synthase subunit 1; diphthamide biosynthesis protein 2 homolog-like 1; diphtheria toxin resistance protein 1; diptheria toxin resistance protein required for diphthamide biosynthesis (Saccharomyces)-like 1; diptheria toxin resistance protein required for diphthamide biosynthesis-like 1; DPH1; DPH1 homolog; DPH1 homolog (S. cerevisiae); DPH1-OVCA2; DPH2L; Dph2l1; DPH2-like 1; DPH-like 1; FLJ33211; hypothetical protein LOC550559; mmDph1; ovarian cancer-associated gene 1 protein; Ovarian cancer-associated gene 1 protein homolog; ovarian tumor suppressor candidate 1; Ovca1; RGD1562694; S-adenosyl-L-methionine:L-histidine 3-amino-3-carboxypropyltransferase 1; Unknown (protein for MGC:127689); zgc:110702
Gene Symbols DPH1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human OVCA1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1801
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.