Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OVCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531447 Supplier Invitrogen™ Supplier No. PA531447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Chromosome 17 is a hotspot for chromosomal aberrations in breast and ovarian cancer. Candidate oncogenes and tumor suppressors located on this chromosome include OVCA1 and OVCA2, p53, BRCA1, HER2/neu and others. OVCA1 and 2 map to a highly conserved region on human chromosome 17p13.3 that is deleted in 80% of ovarian cancers and shows frequent loss of heterozygosity in breast cancers. OVCA1 is induced by BRCA1 and is closely linked to p53, a well-known tumor suppressor gene. The close linkage of OVCA1 and p53 suggests that coordinated loss of the two genes may lead to ovarian, breast and other tumor types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OVCA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene OVCA2
Gene Accession No. Q8WZ82
Gene Alias 9030414M07Rik; candidate tumor suppressor in ovarian cancer 2; candidate tumor suppressor OVCA2; esterase OVCA2; hypothetical protein LOC534099; ortholog of human candidate tumor suppressor in ovarian cancer 2 OVCA2; ovarian cancer gene-2 protein; ovarian cancer-associated gene 2 protein; Ovarian cancer-associated gene 2 protein homolog; ovarian tumor suppressor candidate 2; Ovca2; OVCA2 serine hydrolase domain containing; OVCA2, serine hydrolase domain containing; RGD1564623
Gene Symbols OVCA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human OVCA2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124641
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.