Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OVCH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571233
Description
This target displays homology in the following species: Human: 100%; Rabbit: 79%.
OVCH2 gene ontology annotations related to this gene include serine-type endopeptidase activity.
Specifications
OVCH2 | |
Polyclonal | |
Unconjugated | |
OVCH2 | |
OVCH2; oviductin protease; ovochymase 2; ovochymase 2 (gene/pseudogene); ovochymase-2; OVTN | |
Rabbit | |
Affinity chromatography | |
RUO | |
341277 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7RTZ1 | |
OVCH2 | |
synthetic peptide directed towards the C-terminal region of human OVCH2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction