Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Oven Shroud
SDP

Catalog No. p-7199252 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
120 V
220 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-611-275 120 V
06-611-276 220 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-275 Supplier Restek™ Supplier No. 27186
Only null left
Add to Cart
Add to Cart

Includes Heating Coil and Oven Temperature Sensor

  • Available in 120 V and 220 V units
  • For Agilent 6890/7890 GCs

Specifications

Type 120 V
Includes Heating Coil and Oven Temperature Sensor
For Use With (Application) Agilent 6890/7890 GCs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.