Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OVGP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564007
Description
Immunogen sequence: GTHPRMGNLG LQMEAENRMM LSSSPVIQLP EQTPLAFDNR FVPIYGNHSS VNSVTPQTSP LSLKKEIPEN SAVDE Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 28%.
Oviductins belong to a family of glycoproteins that have been suggested to play several roles during the early processes of reproduction. OVGP1 is a large carbohydrate-rich, epithelial glycoprotein with numerous O-glycosylation sites within threonine, serine, and proline-rich tandem repeats. The protein is secreted from non-ciliated oviductal epithelial cells and associates with ovulated oocytes, blastomeres, and spermatozoan acrosomal regions. At least two isoforms of OVGP are known to exist.
Specifications
OVGP1 | |
Polyclonal | |
Unconjugated | |
OVGP1 | |
120kDa; AU016433; AU019448; CHIT5; chitinase 5; EGP; Estrogen-dependent oviduct protein; MOGP; MUC9; mucin 9; mucin-9; OGP; Ovgp1; oviduct glycoprotein; oviduct glycoprotein OGP; Oviductal glycoprotein; oviductal glycoprotein 1; oviductal glycoprotein 1, 120kD; oviductal glycoprotein 1, 120kDa; oviductin; oviduct-specific glycoprotein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5016 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12889 | |
OVGP1 | |
Recombinant protein corresponding to Human OVGP1. Recombinant protein control fragment (Product #RP-94039). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction