Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OVOL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101057 Supplier Invitrogen™ Supplier No. PA5101057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OVOL1.

OVOL1 is a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.This gene encodes a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OVOL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OVOL1
Gene Accession No. O14753, Q9WTJ2
Gene Alias BB147136; CD254; hOvo1; hRANKL2; Ly109l; movo1; mOvo1a; ODF; OPG; OPG ligand; OPGL; OPTB2; osteoclast differentiation factor; Osteoprotegerin ligand; OVO homolog-like 1; OVO homolog-like 1 (Drosophila); ovo like transcriptional repressor 1; ovo like zinc finger 1; Ovo1; Ovol1; OVO-like 1 binding protein; ovo-like 1(Drosophila); ovo-like zinc finger 1; Putative transcription factor Ovo-like 1; RANKL; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; receptor activator of nuclear factor kappa-B ligand; sOdf; TNF-related activation-induced cytokine; TNFSF11; TNLG6B; Trance; transcription factor Ovo-like 2; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis factor superfamily member 11; tumor necrosis factor-related activation-induced cytokine
Gene Symbols OVOL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OVOL1(Accession O14753), corresponding to amino acid residues M1-E46.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18426, 5017
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.